missing translation for 'onlineSavingsMsg'
Learn More

BU 239 hydrochloride, Tocris Bioscience™

Potent, highly selective I2 ligand

Brand:  Tocris (Bio-Techne) 072650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13457903

  • £337.93 / 50mg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

High affinity ligand for the imidazoline I2 binding site.
Specifications

Chemical Identifiers

1217041-98-9
234.687
bu 239 hydrochloride, 2-4,5-dihydroimidazol-2-yl quinoxaline hydrochloride, 2-4,5-dihydro-1h-imidazol-2-yl quinoxaline hydrochloride
2-(4,5-dihydro-1H-imidazol-2-yl)quinoxaline;hydrochloride
C11H11ClN4
NQMMIEICYUXMKP-UHFFFAOYSA-N
56846046
C1CN=C(N1)C2=NC3=CC=CC=C3N=C2.Cl

Specifications

BU 239 hydrochloride
1217041-98-9
bu 239 hydrochloride, 2-4,5-dihydroimidazol-2-yl quinoxaline hydrochloride, 2-4,5-dihydro-1h-imidazol-2-yl quinoxaline hydrochloride
C1CN=C(N1)C2=NC3=CC=CC=C3N=C2.Cl
234.687
234.69
50mg
C11H11ClN4
NQMMIEICYUXMKP-UHFFFAOYSA-N
2-(4,5-dihydro-1H-imidazol-2-yl)quinoxaline;hydrochloride
56846046
>98%
Videos