Learn More
BU 226 hydrochloride, Tocris Bioscience™
Potent, highly selective I2 ligand
Brand:  Tocris (Bio-Techne) 109110
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13468463
  • £116.13 / 10mg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
A high affinity ligand for I2 imidazoline sites (Ki = 2.7 nM) with very high selectivity (> 2000-fold) over α2-adrenoceptors (Ki = 6700 nM).
Specifications
Chemical Identifiers
1186195-56-1
233.699
bu 226 hydrochloride, 2-4,5-dihydroimidazol-2-yl isoquinoline hydrochloride
3-(4,5-dihydro-1H-imidazol-2-yl)isoquinoline;hydrochloride
C12H12ClN3
NIGHNKWLMSRCOZ-UHFFFAOYSA-N
56972194
C1CN=C(N1)C2=CC3=CC=CC=C3C=N2.Cl
Specifications
10mg
1186195-56-1
bu 226 hydrochloride, 2-4,5-dihydroimidazol-2-yl isoquinoline hydrochloride
C1CN=C(N1)C2=CC3=CC=CC=C3C=N2.Cl
233.699
233.7
BU 226 hydrochloride
C12H12ClN3
NIGHNKWLMSRCOZ-UHFFFAOYSA-N
3-(4,5-dihydro-1H-imidazol-2-yl)isoquinoline;hydrochloride
56972194
>98%