missing translation for 'onlineSavingsMsg'
Learn More

BU 224 hydrochloride, Tocris Bioscience™

Potent, selective I2 ligand. Putative antagonist

Brand:  Tocris (Bio-Techne) 072550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437903

  • £541.68 / 50mg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High affinity ligand for the imidazoline I2 binding site (Ki = 2.1 nM). Putative I2 antagonist; antagonizes the effects of imidazoline ligands on morphine antinociception. Produces ipsiversive rotational behavior in rats with a full 6-OHDA lesion of the nigrostriatal tract.
Specifications

Chemical Identifiers

205437-64-5
233.699
bu224 hydrochloride, bu 224 hydrochloride, 2-4,5-dihydroimidazol-2-yl quinoline hydrochloride, 2-4,5-dihydro-1h-imidazol-2-yl quinoline hydrochloride, bu224 hydrochloride, solid, 2-4,5-dihydroimidazol-2-yl-quinoline hydrochloride, 2-4,5-dihydro-1h-imidazol-2-yl quinoline hydrochloride 1:1
2-(4,5-dihydro-1H-imidazol-2-yl)quinoline;hydrochloride
C12H12ClN3
DDFHQXAQWZWRSQ-UHFFFAOYSA-N
11957470
C1CN=C(N1)C2=NC3=CC=CC=C3C=C2.Cl

Specifications

BU 224 hydrochloride
205437-64-5
bu224 hydrochloride, bu 224 hydrochloride, 2-4,5-dihydroimidazol-2-yl quinoline hydrochloride, 2-4,5-dihydro-1h-imidazol-2-yl quinoline hydrochloride, bu224 hydrochloride, solid, 2-4,5-dihydroimidazol-2-yl-quinoline hydrochloride, 2-4,5-dihydro-1h-imidazol-2-yl quinoline hydrochloride 1:1
C1CN=C(N1)C2=NC3=CC=CC=C3C=C2.Cl
233.699
233.7
50mg
C12H12ClN3
DDFHQXAQWZWRSQ-UHFFFAOYSA-N
2-(4,5-dihydro-1H-imidazol-2-yl)quinoline;hydrochloride
11957470
>98%
Videos
Documents