missing translation for 'onlineSavingsMsg'
Learn More

BTZO 1, Tocris Bioscience™

MIF binder

Brand:  Tocris (Bio-Techne) 420450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13444344

  • £448.52 / 50mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Macrophage migration inhibitory factor (MIF) binder (Kd = 68.6 nM). Activates antioxidant response element (ARE)-mediated gene expression. Suppresses oxidative stress-induced cardiomyocyte apoptosis in vitro. Cardioprotective.
Specifications

Chemical Identifiers

99420-15-2
240.28
btzo 1, 2-2-pyridyl-4h-1,3-benzothiazin-4-one, 2-pyridin-2-yl-4h-benzo e 1,3 thiazin-4-one, 2-pyridin-2-yl-4h-1,3-benzothiazin-4-one, 2-pyridin-2-yl-1,3-benzothiazin-4-one, btzo-1, btzo hplc, 2-2-pyridinyl-4h-1,3-benzothiazin-4-one
2-pyridin-2-yl-1,3-benzothiazin-4-one
C13H8N2OS
GBAKVEWPYUIGHN-UHFFFAOYSA-N
9837640
C1=CC=C2C(=C1)C(=O)N=C(S2)C3=CC=CC=N3

Specifications

BTZO 1
99420-15-2
btzo 1, 2-2-pyridyl-4h-1,3-benzothiazin-4-one, 2-pyridin-2-yl-4h-benzo e 1,3 thiazin-4-one, 2-pyridin-2-yl-4h-1,3-benzothiazin-4-one, 2-pyridin-2-yl-1,3-benzothiazin-4-one, btzo-1, btzo hplc, 2-2-pyridinyl-4h-1,3-benzothiazin-4-one
C1=CC=C2C(=C1)C(=O)N=C(S2)C3=CC=CC=N3
240.28
240.28
50mg
C13H8N2OS
GBAKVEWPYUIGHN-UHFFFAOYSA-N
2-pyridin-2-yl-1,3-benzothiazin-4-one
9837640
>99%
Videos