missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrodes: Handle Only
Handle for use with BTX Two-Needle Array™ Electrodes (cat. no. BTX531).
Brand: BTX™ 450205
Code : NEW
Additional Details : Weight : 0.25000kg
Description
- Reusable
- 45-0167 handle designed for 45-0120 10 mm needle tips
- Used for larger muscle masses
- 45-0168 handle designed for 45-0121 5 mm needle tips
- Recommended for smaller muscle masses
Specifications
In-Vivo Drug or Gene Delivery, Muscle Gene Therapy | |
Delrin | |
Electroporation accessory | |
2-Needle Array Handle, 10 mm | |
3.2 in. | |
10 μsec. to 99 msec. in PBS | |
0.39 in. | |
Non-sterile | |
Square Wave |
2-needle Array (45-0167), ECM830, Gemini X2 and ECM2001+ Generators | |
BTX Electroporation | |
500 V | |
Electrode handle only (requires 45-0167) | |
80 mm | |
1 Ea. | |
10 mm | |
Banana Cables | |
2-needle Array (45-0167) |