missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrodes
Two-Needle Array™ Electrodes designed for in vivo drug or gene delivery.
£268.00
Specifications
For Use With (Application) | In-Vivo Drug or Gene Delivery, Muscle Gene Therapy |
---|---|
For Use With (Equipment) | ECM 830, ECM 2001+, ECM 630, Legacy ECM 2001 Generator |
Voltage | 500 V |
Electrode Shape | Straight/Needle |
Length (English) Electrode | 0.78 in. |
Product Code | Brand | Size (Metric) Gap | Size (English) Gap | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) Gap | Size (English) Gap | Price | Quantity & Availability | ||||
15497260
|
BTX™
450120 |
10 mm | 0.39 in. |
£268.00
Pack of 6 |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15407270
|
BTX™
450121 |
5 mm | 0.19 in. |
£268.00
Pack of 6 |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Sterile, disposable two-needle array
- Enables simple, inexpensive, safe direct injection of muscle tissue for plasmid DNA delivery
- Compatible with ECM™ 830, ECM 2001, ECM 630
Two Sizes
- 5 mm array and handle recommended for small muscle masses such as mouse tibialis
- 10 mm size suitable for larger muscle masses such as rat gastronomes
Specifications
In-Vivo Drug or Gene Delivery, Muscle Gene Therapy | |
500 V | |
0.78 in. | |
10 μsec. to 99 msec. in PBS | |
Sterile | |
Square Wave |
ECM 830, ECM 2001+, ECM 630, Legacy ECM 2001 Generator | |
Straight/Needle | |
20 mm | |
6 Pk. | |
Banana Plug to Banana Plug |
Intended for Research use only. Not for use in diagnostic or clinical procedures.