missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrode Kit
Two-Needle Array™ Electrode Kit designed for in vivo drug or gene delivery.
Brand: BTX™ 450167
Code : NEW
Additional Details : Weight : 0.25000kg
Includes:
- Handle
- Six two-needle arrays
Description
- Consists of reusable array handle and sterile, disposable two-needle array
- Enables simple, inexpensive, safe direct injection of muscle tissue for plasmid DNA delivery
- Compatible with ECM™ 830, ECM 2001, ECM 630
- Supplied in convenient kit with Delrin handle and 20 mm stainless-steel needle array (pack of 6), or order components separately (match handle size to needle array size)
Two Sizes
- 5 mm array and handle recommended for small muscle masses such as mouse tibialis
- 10 mm size suitable for larger muscle masses such as rat gastronomes
Specifications
In-Vivo Drug or Gene Delivery, Muscle Gene Therapy | |
Stainless Steel, Delrin | |
Electroporation accessories | |
2-Needle Array Kit, 10 mm, Pkg. of 6, with Handle | |
6 x 10 mm 2-needle Arrays, 1 x Reusable Handle | |
20 mm | |
5°C to 40°C | |
10 mm | |
Non-sterile | |
Square Wave |
ECM 830, ECM 2001+, ECM 2001, Gemini X2 Generator | |
BTX Electroporation | |
500 V | |
Twin needle | |
0.78 in. | |
10 μsec. to 99 msec. in PBS | |
0.39 in. | |
Needle array | |
Banana Cables |
Intended for Research use only. Not for use in diagnostic or clinical procedures.