missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Microslides for Electrofusion Chambers

Brand:  BTX™ 450105

Code : NEW

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15467250

  • £205.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Allow alignment and fusion to be observed under a microscope
  • Microslide 450 has two stainless steel tubes mounted on a glass slide and generates a divergent field
  • Microslide 453 has two stainless steel bars that can be set apart at 3.2 or 10mm gaps to provide homogenous fields

Specifications

Specifications

Homogeneous
3.2mm
Embryo manipulation, nuclear transfer, cell fusion, hybridoma production, plant protoplast fusion
Stainless Steel
650μL
ECM 830, ECM 2001 generator
Videos