Learn More
BTX™ Genetrodes™ Electrodes
Needle-style reusable Genetrodes™ Electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems.
Brand: BTX™ 450114
Code : NEW
Additional Details : Weight : 0.25000kg
Description
- Needle-style reusable electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Five models available
- Models 508 and 510 are straight with gold tips designed for general in vivo applications
- Models 512, 514, and 516 are L-shaped electrodes with gold tips designed for in vivo and in ovo electroporation
Order Info
Genetrode holder with shaft available separately (Cat. No. BTX515).
Specifications
In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery | |
Gold | |
Genetrodes, 10 mm Straight (GOLD TIP) | |
Genetrode only | |
10 mm | |
1 sec. to 99 msec. | |
1 Ea. | |
1 to 10 mm | |
Non-sterile | |
Square Wave |
ECM 830, EMC 2001, ECM 630 Generator | |
200 V | |
Straight | |
0.39 in. | |
∼1500+ Pulses | |
10 μsec. to 99 msec. in PBS | |
0.03 to 0.39 in. | |
Gold Tip | |
Banana Plug to Micrograbber |
Intended for Research use only. Not for use in diagnostic or clinical procedures.