Learn More
BTX™ Tweezertrode™ Electrode
Tweezertrode™ Electrode is a reusable, tweezer-style, in vivo electrode suitable for drug or gene delivery in animals.
Brand: BTX™ 450118
Code : NEW
Additional Details : Weight : 0.25000kg
Description
- Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel
- Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
- Can be easily cleaned
Order Info
Tweerzertrode kits (Cat. No. BTX450486) and additional cable (BTX524) also available separately.
Specifications
In vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery. | |
Stainless Steel | |
Tweezer Style Electrode | |
Stainless Tweezertrode Electrode, 7 mm Diameter | |
7 mm | |
10 μsec. to 99 msec. in PBS | |
Tweezer Style | |
Non-sterile | |
Square Wave |
ECM 830, ECM 2001+, Gemini X2, and Legacy ECM 2001 | |
BTX Electroporation | |
1000 V | |
0.27 in. | |
Circular | |
1 Ea. | |
11.5 cm | |
Banana to Pin Tip | |
Adaptor Cable |
Intended for Research use only. Not for use in diagnostic or clinical procedures.