missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN2A3P Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549178
Additional Details : Weight : 0.01000kg
Description
BTN2A3P Polyclonal antibody specifically detects BTN2A3P in Human samples. It is validated for Western Blot.Specifications
BTN2A3P | |
Polyclonal | |
Unconjugated | |
BTN2A3P | |
BTN2.3; BTN2A3; BTN2A3P; butyrophilin subfamily 2 member A3, pseudogene | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
54718 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide | |
Q96KV6 | |
BTN2A3P | |
KLH conjugated synthetic peptide between 293-319 amino acids from the Central region of human BTN2A3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |