missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Alfa Aesar BSA Blocking Buffer, 5% in TBS, with 0.05% Tween-20
Brand: Thermo Scientific Alfa Aesar J60098.AP
BSA Blocking Buffer, 5% in TBS, with 0.05% Tween-20
Code : NEW
Additional Details : Weight : 0.00050kg