missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ BSA Blocking Buffer, 5% in PBS, with 0.05% Tween-20

Brand:  Alfa Aesar™ J63711.AK

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15435899

  • POA / 250mL


Estimated Shipment date
from Supplier 10-07-2017
Add to basket

Specifications Description & Specifications

Specifications

Liquid
Blocking Buffer
250mL
−30° to −10°C
Documentation

Technical Documents