Alfa Aesar™ BSA Blocking Buffer, 3% in PBS


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J61655.AP

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.00050kg



Product Code. 15474209

Quantity Price
1 £ 72.19 / 500mL
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Specification

Quantity 500mL
Format Liquid
Storage Requirements −30° to −10°C
Product Type Blocking Buffer
Concentration 3% in PBS