missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ BSA Blocking Buffer, 3% in PBS

Brand:  Alfa Aesar™ J61655.AK

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15464209

  • £41.23 / 250mL


Estimated Shipment date
from Supplier 05-09-2017
Add to basket

Specifications

Specifications

3% in PBS
Blocking Buffer
−30° to −10°C
Liquid
250mL
Documentation

Technical Documents