missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Alfa Aesar BSA Blocking Buffer, 3% in PBS, with 0.05% Tween-20
Brand: Thermo Scientific Alfa Aesar J61116.AP
BSA Blocking Buffer, 3% in PBS, with 0.05% Tween-20
Code : NEW
Additional Details : Weight : 0.00050kg