missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Alfa Aesar BSA Blocking Buffer, 3% in PBS, with 0.02% sodium azide
Brand: Thermo Scientific Alfa Aesar J60473.AP
BSA Blocking Buffer, 3% in PBS, with 0.02% sodium azide
Code : NEW
Additional Details : Weight : 0.00050kg