Learn More
Thermo Scientific Alfa Aesar Bromotris(triphenylphosphine)rhodium(I), 99.95% (metals basis), Rh 10.1% min
Brand: Thermo Scientific Alfa Aesar 039821.01
Bromotris(triphenylphosphine)rhodium(I), 99.95% (metals basis), Rh 10.1% min
Code : NEW
Additional Details : CAS Number : 14973-89-8 Weight : 0.00025kg