missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bromotris(triphenylphosphine)rhodium(I), 99.95% (metals basis), Rh 10.1% min

Hydrogenation

Brand:  Alfa Aesar™ 039821.03

Code : NEW

Additional Details : CAS Number : 14973-89-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11329137

  • £195.90 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

Bromotris(triphenylphosphine)rhodium(I)
14973-89-8
969.69
MFCD00009828

Specifications

Bromotris(triphenylphosphine)rhodium(I)
Crystalline
(metals basis)
MFCD00009828
1g
14973-89-8
99.95%
969.69
Rh 10.1% min.
Soluble in chlorinated solvents
Videos
SDS