missing translation for 'onlineSavingsMsg'
Learn More

Bromotrimethylsilane, 98%, Thermo Scientific Chemicals

Catalog Number p-4509397
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2857-97-8
C3H9BrSi
153.1
IYYIVELXUANFED-UHFFFAOYSA-N
trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
76113
bromo(trimethyl)silane
C[Si](C)(C)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2857-97-8
C3H9BrSi
153.1
IYYIVELXUANFED-UHFFFAOYSA-N
trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromide
76113
bromo(trimethyl)silane
C[Si](C)(C)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Silylation reagent
TRUSTED_SUSTAINABILITY
Melting Point -43.0°C
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 79.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)3SiBr
Refractive Index 1.4200 to 1.4250
Quantity 5 g
Fieser 09,73; 10,59; 12,79; 15,51; 16,50
Specific Gravity 1.16
Solubility Information Solubility in water: decomposes.
Formula Weight 153.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bromotrimethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'