missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromophenol Blue sodium salt, ACS
104.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Bromophenol Blue sodium salt is used as a tracking dye for protein and nucleic acid electrophoresis in polyacrylamide gels. It also used in the preparation of protein samples for western blotting analysis. It finds application in staining histones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Merck Index | 14,1444 |
Solubility Information | Soluble in water. |
Formula Weight | 691.96 |
Grade | ACS |
Quantity | 100 g |
Physical Form | Crystalline |
Chemical Name or Material | Bromophenol Blue sodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'