missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue

Catalog Number 10581334
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
115-39-9
C19H10Br4O5S
669.96
MFCD00005875
UDSAIICHUKSCKT-UHFFFAOYSA-N
3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
8272
CHEBI:59424
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
This item is not returnable. View return policy
115-39-9
C19H10Br4O5S
669.96
MFCD00005875
UDSAIICHUKSCKT-UHFFFAOYSA-N
3',3'',5',5''-Tetrabromophenolsulfonephthalein, BPB, 3', 3'', 5'
8272
CHEBI:59424
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1,lambda{6}-benzoxathiol-3-yl]phenol
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Melting Point 279.0°C
Clarity Passes Test
Beilstein 19, I, 649
Merck Index 15, 1454
Infrared Spectrum Authentic
Formula Weight 669.96
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 3.0 (yellow) to pH 4.6 (blue)
Color Orange/Pink to Red or Purple
Quantity 50 g
Physical Form Powder
Chemical Name or Material Bromophenol Blue, Pure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'