missing translation for 'onlineSavingsMsg'
Learn More

beta-Bromophenetole, 98%, Thermo Scientific™

Catalog Number 10560512
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
589-10-6
C8H9BrO
201.06
MFCD00000234
JJFOBACUIRKUPN-UHFFFAOYSA-N
2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene
68526
2-bromoethoxybenzene
C1=CC=C(C=C1)OCCBr
This item is not returnable. View return policy
589-10-6
C8H9BrO
201.06
MFCD00000234
JJFOBACUIRKUPN-UHFFFAOYSA-N
2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene
68526
2-bromoethoxybenzene
C1=CC=C(C=C1)OCCBr
CAS Min % 97.5
CAS Max % 100.0
Melting Point 33.5°C to 34.0°C
Color Colorless
Boiling Point 144.0°C (40.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OCH2CH2Br
Refractive Index 1.5550 to 1.5570
Quantity 5 g
Beilstein 06, 142
Formula Weight 201.06
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material β-Bromophenetole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • ß-Bromophenetole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'