missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromopentamethylbenzene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Bromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Bromopentamethylbenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | 161°C to 162°C |
Quantity | 25 g |
Beilstein | 1863555 |
Solubility Information | Insoluble in water. |
Formula Weight | 227.15 |
Percent Purity | ≥98% |
Chemical Name or Material | Bromopentamethylbenzene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'