missing translation for 'onlineSavingsMsg'
Learn More

Bromopentacarbonylrhenium(I), 98%, Thermo Scientific Chemicals

Catalog Number 15491777
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This is used to study π-electron deficient complexes between tricarbonylhalorhenium and diimine ligands and also to prepare [3]-ferrocenophane complexes. The starting material for rhenium-polypyridine complexes that have potential applications in catalysis and solar energy conversion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽90°C (Sublimation)
CAS 14220-21-4
Color White
Physical Form Crystalline
Quantity 1 g
Molecular Formula C5BrO5Re
Linear Formula Re(CO)5Br
MDL Number MFCD00013295
UN Number UN3466
Synonym bromopentacarbonylrhenium, bromopentacarbonylrhenium i, bromorhenium; carbon monoxide, pentacarbonylbromorhenium, rebr co 5, bromopentacar-bonylrhenium, pentacarbonylrhenium bromide, rhenium i pentacarbonyl bromide, rhenium,bromopentacarbonyl-, oc-6-22-9ci
Solubility Information Insoluble in water.
InChI Key NWJBOTGGBYFKEJ-UHFFFAOYSA-M
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].Br[Re]
IUPAC Name bromorhenium;carbon monoxide
Molecular Weight (g/mol) 406.161
PubChem CID 6096983
Formula Weight 406.16
Percent Purity 98%
Sensitivity Air & Moisture Sensitive
Chemical Name or Material Bromopentacarbonylrhenium(I)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'