missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Catalog Number p-3757089
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1034-49-7
C19H17Br2P
436.13
MFCD00011864
YFTMLUSIDVFTKU-UHFFFAOYSA-M
bromomethyl triphenylphosphonium bromide, bromomethyltriphenylphosphonium bromide, bromomethyl triphenylphosphoniumbromide, phosphonium, bromomethyl triphenyl-, bromide, bromomethyl triphenylphosphanium bromide, zlchem 395, bromomethyltriphenyl phosphonium bromide, bromomethyl triphenylphosphonium, bromomethyl triphenyl phosphanium bromide
2733422
bromomethyl(triphenyl)phosphanium;bromide
[Br-].BrC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1034-49-7
C19H17Br2P
436.13
MFCD00011864
YFTMLUSIDVFTKU-UHFFFAOYSA-M
bromomethyl triphenylphosphonium bromide, bromomethyltriphenylphosphonium bromide, bromomethyl triphenylphosphoniumbromide, phosphonium, bromomethyl triphenyl-, bromide, bromomethyl triphenylphosphanium bromide, zlchem 395, bromomethyltriphenyl phosphonium bromide, bromomethyl triphenylphosphonium, bromomethyl triphenyl phosphanium bromide
2733422
bromomethyl(triphenyl)phosphanium;bromide
[Br-].BrC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 230.0°C to 235.0°C
Color Beige to Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula BrCH2P(C6H5)3Br
Quantity 5 g
Formula Weight 436.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (Bromomethyl)triphenylphosphonium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'