missing translation for 'onlineSavingsMsg'
Learn More

Bromoethane, ≥99%, extra pure, Acros Organics

Brand:  Acros Organics 330350050

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 74-96-4 Weight : 0.06000kg Transport : UN number : 1891 Chem class : 1 Pack group : II

Quantity 5mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10173381

  • £34.90 / 5mL


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

Bromoethane
74-96-4
≥99.0% (GC)
C2H5Br
108.97
Ethyl bromide
99+%
Extra Pure
99+%
C2H5Br
MFCD00000232

Specifications

Bromoethane
0.0005 % max. (as HBr)
Authentic
1.4230 to 1.4250 (20°C, 589nm)
White to beige to light brown
≥99.0% (GC)
5mL
C2H5Br
MFCD00000232
99+%
74-96-4
Glass bottle
Extra Pure
Powder and/or Crystals
99+%
C2H5Br
108.97
Ethyl bromide
Documentation

Technical Documents