Bromoethane, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 154215000

500ML Bromoethane, 98%, pure

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 74-96-4 Weight: 0.78000kg Transport: UN number : 1891 Chem class : 1 Pack group : II


Packaging Glass bottle
Quantity 500mL

Product Code. 10173250

Quantity Price
1 £ 18.5 / 500mL
EU Stock 11
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Bromoethane
Name Note Pure
CAS 74-96-4
Purity Grade Extra Pure
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2H5Br
Linear Formula C2H5Br
Formula Weight 108.97
MDL Number MFCD00000232
Synonym Ethyl bromide

Specification

Assay ≥97.5%
Physical Form Liquid
Formula Weight 108.97
Quantity 500mL
Color Clear colorless to slightly yellow
Linear Formula C2H5Br
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Purity Grade Extra Pure
CAS 74-96-4
Infrared Spectrum Authentic
Refractive Index 1.4230 to 1.4250 (20°C, 589nm)
Assay Percent Range 98%
MDL Number MFCD00000232
Packaging Glass bottle
Water 0.03% max.
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
Name Note Pure
Molecular Formula C2H5Br
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Bromoethane
Merck Index 15, 3826
Synonym Ethyl bromide
Beilstein 01, 88
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.