missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bromocyclohexane, 99%

Brand:  Alfa Aesar™ 031031.22

Code : NEW

Additional Details : CAS Number : 108-85-0 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11317388

  • £20.56 / 100g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Bromocyclohexane
C6H11Br
MFCD00003819
14,2730
108-85-0
163.06
1098373
Cyclohexyl bromide

Specifications

Bromocyclohexane
1.4950
99%
C6H11Br
1.335g/mL
1098373
−57°C
166° to 167°C
62° (143°F)
108-85-0
Liquid
99%
163.06
MFCD00003819
14,2730
Cyclohexyl bromide
100g
Air & Light Sensitive
Videos
SDS