Learn More
Bromocresol Purple, Pure, Indicator Grade, Solid, Fisher Chemical™
C21H16Br2O5S, CAS Number-115-40-2, 25g, Red, 540.21g/mol, 245 deg.C, 11681, Glass Jar, Powder Solid
Brand: Fisher Chemical B/4330/44
Code : 21
Additional Details : CAS Number : 115-40-2 Weight : 0.00500kg
Description
C21H16Br2O5S, CAS Number-115-40-2, 25g, Red, 540.21g/mol, 245 deg.C, 11681, Glass Jar, Powder Solid
Specifications
245°C | |
C21H16Br2O5S | |
540.21g/mol | |
Red | |
Powder |
115-40-2 | |
11681 | |
Glass Jar | |
5 g |