missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Bromocresol Green, pure, indicator grade
32.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Melting Point | 225°C, 437°F |
Merck Index | 15, 1393 |
Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in benzene, ethanol (30g/L), very soluble in diethyl ether and ethyl acetate |
Infrared Spectrum | Authentic |
Formula Weight | 698.01 |
Grade | Pure |
Packaging | Glass bottle |
Specific Extinction | 600 min. (617nm, in buffer) |
Visual Transition Interval | from pH 3.8 (yellow) to pH 5.4 (blue) |
Color | Brown |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'