missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromobenzene, 99%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -31.0°C |
Color | Colorless to Yellow |
Density | 1.4900g/mL |
Boiling Point | 156.0°C |
Flash Point | 51°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Plastic Drum |
Linear Formula | C6H5Br |
Refractive Index | 1.5580 to 1.5600 |
Show More |
Safety and Handling
Product Identifier
- Bromobenzene
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362-Take off contaminated clothing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'