missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-4089880
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
22118-09-8
C2H2BrClO
157.39
MFCD00000724
SYZRZLUNWVNNNV-UHFFFAOYSA-N
bromoacetyl chloride, acetyl chloride, bromo, bromo-acetyl chloride, monobromoacetic acid chloride, bromoacetylchloride, bromacetylchioride, bromacetylchloride, bromoacetylchioride, bromacetyl chloride, bromo-acetylchloride
89602
2-bromoacetyl chloride
C(C(=O)Cl)Br
22118-09-8
C2H2BrClO
157.39
MFCD00000724
SYZRZLUNWVNNNV-UHFFFAOYSA-N
bromoacetyl chloride, acetyl chloride, bromo, bromo-acetyl chloride, monobromoacetic acid chloride, bromoacetylchloride, bromacetylchioride, bromacetylchloride, bromoacetylchioride, bromacetyl chloride, bromo-acetylchloride
89602
2-bromoacetyl chloride
C(C(=O)Cl)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Density 1.9000g/mL
Boiling Point 127.0°C to 128.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2COCl
Refractive Index 1.496
Quantity 25 g
Beilstein 02,215
Specific Gravity 1.9
Solubility Information Solubility in water: reacts
Formula Weight 157.39
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Bromoacetyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'