missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetyl bromide, 98%, Thermo Scientific Chemicals

Catalog Number p-7024612
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
598-21-0
C2H2Br2O
201.845
MFCD00000115
LSTRKXWIZZZYAS-UHFFFAOYSA-N
bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide
61147
2-bromoacetyl bromide
C(C(=O)Br)Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
598-21-0
C2H2Br2O
201.845
MFCD00000115
LSTRKXWIZZZYAS-UHFFFAOYSA-N
bromoacetyl bromide, acetyl bromide, bromo, bromoacetylbromide, acetyl bromide, 2-bromo, bromo acetyl bromide, ccris 9090, bromoacetic acid bromide, bromacetylbromide, bromacetyl bromide, bromoactyl bromide
61147
2-bromoacetyl bromide
C(C(=O)Br)Br

Bromoacetyl bromide is widely used in the synthesis of fine chemicals, agrochemicals, dyes and pharmaceuticals. As an acylating agent, it reacts with 10H-phenothiazine to give 10-bromoacetyl-10H-phenothiazine. Similarly, it is also involved in the preparation of bromoactylated starch which is a biodegradable thermoplastic copolymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetyl bromide is widely used in the synthesis of fine chemicals, agrochemicals, dyes and pharmaceuticals. As an acylating agent, it reacts with 10H-phenothiazine to give 10-bromoacetyl-10H-phenothiazine. Similarly, it is also involved in the preparation of bromoactylated starch which is a biodegradable thermoplastic copolymer.

Solubility
Immiscible with water.

Notes
Moisture Sensitive. Store in cool place. Hydrolyses readily. Incompatible with strong oxidizing agents, alcohols and strong bases.
TRUSTED_SUSTAINABILITY
Density 2.324
Boiling Point 147°C to 150°C
Flash Point >105°C (221°F)
Odor Acrid
Refractive Index 1.546
Quantity 25 g
UN Number UN2513
Beilstein 605440
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 201.86
Percent Purity 98%
Chemical Name or Material Bromoacetyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'