missing translation for 'onlineSavingsMsg'
Learn More

α'-Bromo-α,α,α-trifluoro-p-xylene, 98%, Thermo Scientific Chemicals

Catalog Number p-77126
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
402-49-3
C8H6BrF3
239.04
MFCD00000403
IKSNDOVDVVPSMA-UHFFFAOYSA-N
4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene
123062
1-(bromomethyl)-4-(trifluoromethyl)benzene
C1=CC(=CC=C1CBr)C(F)(F)F
This item is not returnable. View return policy
402-49-3
C8H6BrF3
239.04
MFCD00000403
IKSNDOVDVVPSMA-UHFFFAOYSA-N
4-trifluoromethyl benzyl bromide, 1-bromomethyl-4-trifluoromethyl benzene, 4-trifluoromethylbenzyl bromide, 4-bromomethyl benzotrifluoride, p-trifluoromethylbenzyl bromide, 4-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-trifluoromethyl, 4-trifluoromethylbenzylbromide, p-trifluoromethyl benzyl bromide, alpha'-bromo-alpha,alpha,alpha-trifluoro-p-xylene
123062
1-(bromomethyl)-4-(trifluoromethyl)benzene
C1=CC(=CC=C1CBr)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 30°C to 34°C
Color White to Yellow
Density 1.5400g/mL
Boiling Point 65°C to 69°C (5.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4CH2Br
Quantity 1 g
Specific Gravity 1.54
Formula Weight 239.04
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material α'-Bromo-α, α, α-trifluoro-p-xylene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'