missing translation for 'onlineSavingsMsg'
Learn More

α-Bromo-o-tolunitrile, 98%, ACROS Organics™

Brand:  Acros Organics 163135000

Code : 29

Additional Details : CAS Number : 22115-41-9 Weight : 0.55000kg Transport : UN number : 1759 Chem class : 1a Pack group : II


Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10761614

  • £783.30 / 500g


Estimated Shipment
17-05-2018
Add to basket


Specifications

Chemical Identifiers

α-Bromo-o-tolunitrile
97.5
≥97.5%
C8H6BrN
196.05
22115-41-9
100.0
98%
BrCH2C6H4CN
MFCD00001794

Specifications

α-Bromo-o-tolunitrile
97.5
Authentic
Crystalline Powder or Chunks
98%
BrCH2C6H4CN
MFCD00001794
White to gray-brown
2-Cyanobenzyl bromide
22115-41-9
100.0
Glass bottle
≥97.5%
C8H6BrN
196.05
09, 470
70° to 74°C
500g
Videos
SDS