missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-2'-acetonaphthone, 98%, ACROS Organics™

£29.09 - £118.97

Chemical Identifiers

CAS 613-54-7
Molecular Formula C12H9BrO
Molecular Weight (g/mol) 249.11
MDL Number MFCD00004109
InChI Key YHXHHGDUANVQHE-UHFFFAOYSA-N
Synonym 2-bromo-2'-acetonaphthone, bromomethyl 2-naphthyl ketone, 2-bromoacetyl naphthalene, 2-bromo-1-naphthalen-2-yl ethanone, 2-bromo-1-2-naphthyl-1-ethanone, ethanone, 2-bromo-1-2-naphthalenyl, 2-bromo-1-2-naphthyl ethanone, 2-bromo-1-naphthalen-2-yl ethan-1-one, alpha-bromo-2'-acetonaphthone, 2-bromo-1-naphthalen-2-yl-ethanone
PubChem CID 69179
IUPAC Name 2-bromo-1-naphthalen-2-ylethanone
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)CBr
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10346740
SDS View Documents
Acros Organics
147480100
10g £29.09
10g View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
10031650
SDS View Documents
Acros Organics
147480500
50g £118.97
50g View alternative products
Save Up to 
»
Estimated Shipment
14-12-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

613-54-7
249.11
YHXHHGDUANVQHE-UHFFFAOYSA-N
69179
C1=CC=C2C=C(C=CC2=C1)C(=O)CBr
C12H9BrO
MFCD00004109
2-bromo-2'-acetonaphthone, bromomethyl 2-naphthyl ketone, 2-bromoacetyl naphthalene, 2-bromo-1-naphthalen-2-yl ethanone, 2-bromo-1-2-naphthyl-1-ethanone, ethanone, 2-bromo-1-2-naphthalenyl, 2-bromo-1-2-naphthyl ethanone, 2-bromo-1-naphthalen-2-yl ethan-1-one, alpha-bromo-2'-acetonaphthone, 2-bromo-1-naphthalen-2-yl-ethanone
2-bromo-1-naphthalen-2-ylethanone

Specifications

α-Bromo-2'-acetonaphthone
Glass bottle
82°C to 85°C
98%
97.5
97.5% min. (GC)
C10H7COCH2Br
2-bromo-2'-acetonaphthone, bromomethyl 2-naphthyl ketone, 2-bromoacetyl naphthalene, 2-bromo-1-naphthalen-2-yl ethanone, 2-bromo-1-2-naphthyl-1-ethanone, ethanone, 2-bromo-1-2-naphthalenyl, 2-bromo-1-2-naphthyl ethanone, 2-bromo-1-naphthalen-2-yl ethan-1-one, alpha-bromo-2'-acetonaphthone, 2-bromo-1-naphthalen-2-yl-ethanone
C1=CC=C2C=C(C=CC2=C1)C(=O)CBr
249.11
249.11
98%
Authentic
Brown-Purple
10g
613-54-7
100.0
C12H9BrO
MFCD00004109
YHXHHGDUANVQHE-UHFFFAOYSA-N
2-bromo-1-naphthalen-2-ylethanone
69179
Crystalline Powder
Safety and Handling

Safety and Handling

Product Identifier
  • a-Bromo-2'-acetonaphthone

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.