missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzyl bromide, 99+%, Thermo Scientific™

Catalog Number p-73714
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
20443-98-5
C7H5BrCl2
239.92
MFCD00000577
PDFGFQUSSYSWNI-UHFFFAOYSA-N
2,6-dichlorobenzyl bromide, 2-bromomethyl-1,3-dichlorobenzene, alpha-bromo-2,6-dichlorotoluene, 2,6-dichlorobenzylbromide, benzene, 2-bromomethyl-1,3-dichloro, 1-bromomethyl-2,6-dichlorobenzene, .alpha.-bromo-2,6-dichlorotoluene, a-bromo-2,6-dichlorotoluene, 2,6 dichloro benzyl bromide, benzene, 2-bromomethyl-1,3-dichloro-9ci
30159
2-(bromomethyl)-1,3-dichlorobenzene
ClC1=CC=CC(Cl)=C1CBr
This item is not returnable. View return policy
20443-98-5
C7H5BrCl2
239.92
MFCD00000577
PDFGFQUSSYSWNI-UHFFFAOYSA-N
2,6-dichlorobenzyl bromide, 2-bromomethyl-1,3-dichlorobenzene, alpha-bromo-2,6-dichlorotoluene, 2,6-dichlorobenzylbromide, benzene, 2-bromomethyl-1,3-dichloro, 1-bromomethyl-2,6-dichlorobenzene, .alpha.-bromo-2,6-dichlorotoluene, a-bromo-2,6-dichlorotoluene, 2,6 dichloro benzyl bromide, benzene, 2-bromomethyl-1,3-dichloro-9ci
30159
2-(bromomethyl)-1,3-dichlorobenzene
ClC1=CC=CC(Cl)=C1CBr

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 54°C to 57°C
Color Colorless to White
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula Cl2C6H3CH2Br
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: 1g/10mL methanol
Formula Weight 239.93
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 6-Dichlorobenzyl bromide, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dichlorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'