Learn More
Bromine, 1M solution in trimethyl phosphate, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 428721000
Code : 29
Additional Details : Weight : 0.13070kg Transport : UN number : 1744 Chem class : 8 Pack group : I
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
58.7°C | |
-7.2°C | |
7726-95-6,512-56-1 | |
Br | |
113°C | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
[Br-] | |
79.91 | |
CHEBI:29224 | |
ACS Reagent | |
1.307 |
Bromine | |
1M Solution in trimethyl Phosphate | |
100 mL | |
Br2 | |
15,1401 | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
molecular bromine | |
24408 | |
159.8 | |
AcroSeal™ Glass Bottle | |
1.3070g/mL |
Safety and Handling
- Bromine, 1M solution in trimethyl phosphate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1B
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H340-May cause genetic defects.
- H351-Suspected of causing cancer.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only