missing translation for 'onlineSavingsMsg'
Learn More

Bromine, 99+%, extra pure, ACROS Organics™

Brand:  Acros Organics 402845000

Code : 29

Additional Details : CAS Number : 7726-95-6 Weight : 1.60550kg Transport : UN number : 1744 Chem class : 1a Pack group : I


Quantity 500mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10483683

  • £65.80 / 500mL


In Stock 25
EU Stock 4
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

Bromine
Extra Pure
99+%
Br2
MFCD00010896
7726-95-6
≥99%
Br2
159.82

Specifications

Bromine
Extra Pure
≥99%
Br2
159.82
03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47
500ppm max.
2ppm max.
Glass bottle
3.111
58.7°C
− 7.2°C
7726-95-6
Fuming Liquid
99+%
Br2
MFCD00010896
15, 1401
3.1110g/mL
1ppm max.
Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl, aq solns of bromides, freely soluble in alcohol, ether, chcl3, ccl4, cs2
0.314 mm2/s (25°C)
Red-brown
500mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bromine

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS