missing translation for 'onlineSavingsMsg'
Learn More

Bromine, 99.6%, for analysis, ACROS Organics™

Brand:  Acros Organics 196662500

38.94 GBP valid until 2021-07-31
Use promo code "16115" to get your promotional price.



Code : 29

Additional Details : CAS Number : 7726-95-6 Weight : 0.83000kg Transport : UN number : 1744 Chem class : 8 Pack group : I


Quantity 250mL

 View more versions of this product

Product Code. 10062600

  • £53.81 / 250mL

In stock, ready to ship: 114

Please to purchase this item. Need a web account? Register with us today!


Sea freight only: For EU stock allow up to 15 days delivery.

Description

Description

C-X bond formation reagent for analysis
Specifications

Chemical Identifiers

7726-95-6
159.82
GDTBXPJZTBHREO-UHFFFAOYSA-N
24408
molecular bromine
Br2
MFCD00010896
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
CHEBI:29224
BrBr

Specifications

Bromine
7726-95-6
Br2
MFCD00010896
15, 1401
Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl, aq solns of bromides, freely soluble in alcohol, ether, chcl3, ccl4, cs2
BrBr
159.82
CHEBI:29224
99.6%
3.1110g/mL
Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl, aq solns of bromides, freely soluble in alcohol, ether, chcl3, ccl4, cs2
0.314 mm2/s (25°C)
-7.2°C
p.a.
99.5% min. (Iodometry)
Br2
03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
GDTBXPJZTBHREO-UHFFFAOYSA-N
molecular bromine
24408
159.82
Analysis
Glass bottle
3.111
58.7°C
250mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bromine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.

SDS Documents
Certificates