missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Broad Base Crucibles
£63.55 - £273.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12877743
|
Yorlab
10448/11 |
25 mL |
£273.00
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
12887743
|
Yorlab
10448/12 |
40 mL |
£63.55
Each |
In Stock
Log in to see stock. |
|||||
Specifications
Silica | |
Crucible |