missing translation for 'onlineSavingsMsg'
Learn More

BRL 44408 maleate, Tocris Bioscience™

Selective α2A antagonist

Brand:  Tocris (Bio-Techne) 113350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13488633

  • £539.52 / 50mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Selective α2A-adrenoceptor antagonist (Ki = 1.7 nM and 144.5 nM at α2A and α2B-adrenergic receptors respectively). Increases hippocampal noradrenalin release following systemic administration. Also available as part of the α2-Adrenoceptor Tocriset™.
Specifications

Chemical Identifiers

681806-46-2
331.372
2-[(4, 5-Dihydro-1H-imidazol-2-yl)methyl]-2, 3-dihydro-1-methyl-1H-isoindole maleate
(Z)-but-2-enedioic acid;2-(4,5-dihydro-1H-imidazol-2-ylmethyl)-1-methyl-1,3-dihydroisoindole
C17H21N3O4
DDIQGSUEJOOQQQ-BTJKTKAUSA-N
10382026
CC1C2=CC=CC=C2CN1CC3=NCCN3.C(=CC(=O)O)C(=O)O

Specifications

BRL 44408 maleate
681806-46-2
C17H21N3O4
DDIQGSUEJOOQQQ-BTJKTKAUSA-N
(Z)-but-2-enedioic acid;2-(4,5-dihydro-1H-imidazol-2-ylmethyl)-1-methyl-1,3-dihydroisoindole
10382026
50mg
>98%
2-[(4, 5-Dihydro-1H-imidazol-2-yl)methyl]-2, 3-dihydro-1-methyl-1H-isoindole maleate
CC1C2=CC=CC=C2CN1CC3=NCCN3.C(=CC(=O)O)C(=O)O
331.372
331.37
Videos