Learn More
Grant Instruments™ Grant Optima Bridge Plate for use with Plastic Tanks
Fits Grant G Optima Heating Circulators to a range of plastic tanks
Brand:  Grant Instruments™ G-BPS
Code : 57
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12737799
  • £117.30 / Each
Estimated Shipment
05-07-2018
Add to basket
Description
Description
  • G-BPL: Fits G Optima Heating Ciruclator models to P12 and P18 plastic tanks
  • G-BPS: Fits G Optima Heating Ciruclator models to P5 plastic tank
Specifications
Specifications
Grant G Optima Heating Circulators P-5
Plastic
Videos