Learn More
Greiner Bio-One™ BREATHseal™ Sealer
For the culture of prokaryotic and eukaryotic cells
Brand:  Greiner Bio-One™ 676051
Additional Details : Weight : 0.10000kg
Product Code. 10472353
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Unique gas-permeable sealer with a wide distribution of pores for excellent gas exchange
  • Available sterile and non-sterile