Learn More
eBioscience™ BrdU Kit for IHC/ICC Colorimetric
Contains the necessary reagents and buffers for identifying and examining proliferating cells by immunohistochemical or immunocytochemical analysis. Optimized for IHC with both frozen and paraffin-embedded BrdU-labeled mouse intestine and ICC of BrdU-pulsed HeLa cells grown on culture slides.
Brand:  Affymetrix eBioscience 8800-6599-42
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15501247
  • £317.03 / 100 tests
Estimated Shipment date
from Supplier 26-04-2018
Add to basket
Contains Sodium azide, formaldehyde, and hydrogen peroxide
Includes: BrdU Fixation Buffer
BrdU Antigen Retrieval Solution (10X)
BrdU Blocking Solution
Anti-BrdU Biotin (clone BU20A) : 10 μg
3, 3'-Diaminobenzidine (DAB) Substrate Solution: Reagent A (DAB Chromogen), Reagent B (DAB Diluent), and Dropper Bottle (for storage and application of working DAB solution)
  • Cells are labeled in vitro or in vivo with 5-bromo-2′-deoxyuridine (BrdU), which is incorporated into DNA in place of thymidine during the S-phase of the cell cycle
  • Following fixation and antigen-retrieval steps, cells or tissue sections are stained for BrdU incorporation and visualized using an Avidin-HRP and DAB substrate reaction
Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections, Microscopy, Immunocytochemistry, Immunohistochemical Staining of Frozen Tissue Sections
100 tests
Chimpanzee, Human, Macaque, Mouse, Rat
Refer to individual components