missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Brass spheres, alloy 260, 12.7mm (0.5 in.) dia.

Brand:  Alfa Aesar™ 045184.KM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 25 pieces

Product Code. 15402197

  • £33.92 / Pack of 25


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Specifications

Brass spheres, Alloy 260
25 pieces
12.7mm (0.5 in.) dia.
MFCD00171255
Videos
SDS