missing translation for 'onlineSavingsMsg'
Learn More

Endecotts™ Brass Lid

For use with Endecotts™ sieves. Endecotts™ Brass Lids are widely used with test sieves for all types of laboratory sampling and particle size analysis.

Brand:  Endecotts™ 200BRASLID

Code : 03

Additional Details : Weight : 0.33840kg

 View more versions of this product

Product Code. 12941985

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Brass lid for Endecotts Sieves
  • 200mm
Specifications

Specifications

Lid
200 mm
1 Ea.
Brass
For sieves
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers