missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Brass gauze, alloy 260, 60 mesh woven from 0.19mm (0.0075 in.) dia. wire

Brand:  Thermo Scientific Alfa Aesar 045222.GT

Brass gauze, alloy 260, 60 mesh woven from 0.19mm (0.0075 in.) dia. wire

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15436835

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.