missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Brass gauze, alloy 260, 60 mesh woven from 0.19mm (0.0075 in.) dia. wire

Brand:  Alfa Aesar™ 045222.HB

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300 x 300mm

 View more versions of this product

Product Code. 15446835

  • £19.19 / Each


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

Brass gauze, Alloy 260
60 mesh woven from 0.19mm (0.0075 in.) dia. wire
MFCD00171255

Specifications

Brass gauze, Alloy 260
60 Mesh
60 mesh woven from 0.19mm (0.0075 in.) dia. wire
MFCD00171255
8.53g/mL
300 x 300mm
Odorless
Videos
SDS